Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTVGKSSKM LQHIDYRMRC ILQDGRIFIG TFKAFDKHMN LILCDCDEFR KIKPKNSKQA EREEKRVLGL VLLRGENLVS MTVEGPPPKD TGIARVPLAG AAGGPGIGRA AGRGIPAGVP MPQAPAGLAG PVRGVGGPSQ QVMTPQGRGT VAAAAAAATA SIAGAPTQYP PGRGGPPPPM GRGAPPPGMM GPPPGMRPPM GPPMGIPPGR GTPMGMPPPG MRPPPPGMRG KCLQW (1-234 aa encoded by BC003530)
The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B polypeptide of Sm protein; Sm protein B/B'; Sm-B/B'; sm-B/Sm-B'; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; Small nuclear ribonucleoprotein-associated proteins B and B'; snRNP-B
Gene Aliases: CCMS; COD; Sm-B/B'; SmB/B'; SmB/SmB'; snRNP-B; SNRPB; SNRPB1
UniProt ID: (Human) P14678
Entrez Gene ID: (Human) 6628
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support