Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sm-D autoantigen; Sm-D1; small nuclear ribonucleoprotein D1 polypeptide 16kDa pseudogene; Small nuclear ribonucleoprotein Sm D1; snRNP core protein D1
Gene Aliases: HsT2456; Sm-D1; SMD1; SNRPD; SNRPD1
UniProt ID: (Human) B5BTZ1
Entrez Gene ID: (Human) 6632
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support