Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSKAHPPEL KKFMDKKLSL KLNGGRHVQG ILRGFDPFMN LVIDECVEMA TSGQQNNIGM VVIRGNSIIM LEALERV (1-76 aa encoded by B C000070)
Core component of the spliceosomal U1, U2, U4 and U5 small nuclear ribonucleoproteins (snRNPs), the building blocks of the spliceosome. Thereby, plays an important role in the splicing of cellular pre-mRNAs. Most spliceosomal snRNPs contain a common set of Sm proteins SNRPB, SNRPD1, SNRPD2, SNRPD3, SNRPE, SNRPF and SNRPG that assemble in an heptameric protein ring on the Sm site of the small nuclear RNA to form the core snRNP. Appears to function in the U7 snRNP complex that is involved in histone 3'- end processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sm protein G; Sm-G; Small nuclear ribonucleoprotein G; snRNP-G
Gene Aliases: 2810024K17Rik; AL022803; PBSCG; Sm-G; SMG; snRNP-G; SNRPG
UniProt ID: (Human) P62308, (Mouse) P62309
Entrez Gene ID: (Human) 6637, (Mouse) 68011
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support