Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MFPEQQKEEF VSVWVRDPRI QKEDFWHSYI DYEICIHTNS MCFTMKTSCV RRRYREFVWL RQRLQSNALL VQLPELPSKN LFFNMNNRQH VDQRRQGLED FLRKVLQNAL LLSDSSLHLF LQSHLNSEDI EACVSGQTKY SVEEAIHKFA LMNRRFPEED EEGKKENDID YDSESSSSGL GHSSDDSSSH GCKVNTAPQE S
Probable phosphoinositide-binding protein involved in protein sorting and membrane trafficking in endosomes. Plays a role in cilium biogenesis through regulation of the transport and the localization of proteins to the cilium. Required for the localization to the cilium of V-ATPase subunit ATP6V1D and ATP6V0D1, and RAB8A. Involved in osteoclast differentiation and therefore bone resorption.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sorting nexin-10
Gene Aliases: OPTB8; SNX10
UniProt ID: (Human) Q9Y5X0
Entrez Gene ID: (Human) 29887
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support