Product References
New insights into the phosphorylation of the threonine motif of the β1 integrin cytoplasmic domain.
Life science alliance
Böttcher RT,Strohmeyer N,Aretz J,Fässler R
Fri Apr 01 00:00:00 UTC 2022
Sorting nexin 17 increases low-density lipoprotein receptor-related protein 4 membrane expression: A novel mechanism of acetylcholine receptor aggregation in myasthenia gravis.
Frontiers in immunology
He X,Zhou S,Ji Y,Zhang Y,Lv J,Quan S,Zhang J,Zhao X,Cui W,Li W,Liu P,Zhang L,Shen T,Fang H,Yang J,Zhang Y,Cui X,Zhang Q,Gao F
Fri Nov 04 00:00:00 UTC 2022
Sorting nexin 17 increases low-density lipoprotein receptor-related protein 4 membrane expression: A novel mechanism of acetylcholine receptor aggregation in myasthenia gravis.
Frontiers in immunology
He X,Zhou S,Ji Y,Zhang Y,Lv J,Quan S,Zhang J,Zhao X,Cui W,Li W,Liu P,Zhang L,Shen T,Fang H,Yang J,Zhang Y,Cui X,Zhang Q,Gao F
Fri Nov 04 00:00:00 UTC 2022
Recycling of autophagosomal components from autolysosomes by the recycler complex.
Nature cell biology
Zhou C,Wu Z,Du W,Que H,Wang Y,Ouyang Q,Jian F,Yuan W,Zhao Y,Tian R,Li Y,Chen Y,Gao S,Wong CCL,Rong Y
Fri Apr 01 00:00:00 UTC 2022
Molecular determinants of αVβ5 localization in flat clathrin lattices - role of αVβ5 in cell adhesion and proliferation.
Journal of cell science
Zuidema A,Wang W,Kreft M,Bleijerveld OB,Hoekman L,Aretz J,Böttcher RT,Fässler R,Sonnenberg A
Wed Jun 01 00:00:00 UTC 2022
The Role of Sorting Nexin 17 in Cardiac Development.
Frontiers in cardiovascular medicine
Wu Y,Zhou Y,Huang J,Ma K,Yuan T,Jiang Y,Ye M,Li J
Fri Jan 07 00:00:00 UTC 2022
A genome-wide CRISPR screen identifies host factors that regulate SARS-CoV-2 entry.
Nature communications
Zhu Y,Feng F,Hu G,Wang Y,Yu Y,Zhu Y,Xu W,Cai X,Sun Z,Han W,Ye R,Qu D,Ding Q,Huang X,Chen H,Xu W,Xie Y,Cai Q,Yuan Z,Zhang R
Thu Feb 11 00:00:00 UTC 2021
SNX17 protects the heart from doxorubicin-induced cardiotoxicity by modulating LMOD2 degradation.
Pharmacological research
Zhang Y,Ni L,Lin B,Hu L,Lin Z,Yang J,Wang J,Ma H,Liu Y,Yang J,Lin J,Xu L,Wu L,Shi D
Thu Jul 01 00:00:00 UTC 2021
Suppression of Membranous LRP5 Recycling, Wnt/β-Catenin Signaling, and Colon Carcinogenesis by 15-LOX-1 Peroxidation of Linoleic Acid in PI3P.
Cell reports
Liu F,Zuo X,Liu Y,Deguchi Y,Moussalli MJ,Chen W,Yang P,Wei B,Tan L,Lorenzi PL,Gao S,Jaoude JC,Mehdizadeh A,Valentin LA,Wei D,Shureiqi I
Tue Aug 18 00:00:00 UTC 2020
Suppression of Membranous LRP5 Recycling, Wnt/β-Catenin Signaling, and Colon Carcinogenesis by 15-LOX-1 Peroxidation of Linoleic Acid in PI3P.
Cell reports
Liu F,Zuo X,Liu Y,Deguchi Y,Moussalli MJ,Chen W,Yang P,Wei B,Tan L,Lorenzi PL,Gao S,Jaoude JC,Mehdizadeh A,Valentin LA,Wei D,Shureiqi I
Tue Aug 18 00:00:00 UTC 2020
TFEB controls retromer expression in response to nutrient availability.
The Journal of cell biology
Curnock R,Calcagni A,Ballabio A,Cullen PJ
Mon Dec 02 00:00:00 UTC 2019
Retriever is a multiprotein complex for retromer-independent endosomal cargo recycling.
Nature cell biology
McNally KE,Faulkner R,Steinberg F,Gallon M,Ghai R,Pim D,Langton P,Pearson N,Danson CM,Nägele H,Morris LL,Singla A,Overlee BL,Heesom KJ,Sessions R,Banks L,Collins BM,Berger I,Billadeau DD,Burstein E,Cullen PJ
Sun Oct 01 00:00:00 UTC 2017
Retriever is a multiprotein complex for retromer-independent endosomal cargo recycling.
Nature cell biology
McNally KE,Faulkner R,Steinberg F,Gallon M,Ghai R,Pim D,Langton P,Pearson N,Danson CM,Nägele H,Morris LL,Singla A,Overlee BL,Heesom KJ,Sessions R,Banks L,Collins BM,Berger I,Billadeau DD,Burstein E,Cullen PJ
Sun Oct 01 00:00:00 UTC 2017
A phosphoinositide conversion mechanism for exit from endosomes.
Nature
Ketel K,Krauss M,Nicot AS,Puchkov D,Wieffer M,Müller R,Subramanian D,Schultz C,Laporte J,Haucke V
Thu Jan 21 00:00:00 UTC 2016
A phosphoinositide conversion mechanism for exit from endosomes.
Nature
Ketel K,Krauss M,Nicot AS,Puchkov D,Wieffer M,Müller R,Subramanian D,Schultz C,Laporte J,Haucke V
Thu Jan 21 00:00:00 UTC 2016
Regulation of Cell Migration and β1 Integrin Trafficking by the Endosomal Adaptor GGA3.
Traffic (Copenhagen, Denmark)
Ratcliffe CD,Sahgal P,Parachoniak CA,Ivaska J,Park M
Wed Jun 01 00:00:00 UTC 2016
Regulation of Cell Migration and β1 Integrin Trafficking by the Endosomal Adaptor GGA3.
Traffic (Copenhagen, Denmark)
Ratcliffe CD,Sahgal P,Parachoniak CA,Ivaska J,Park M
Wed Jun 01 00:00:00 UTC 2016
Regulation of Cell Migration and β1 Integrin Trafficking by the Endosomal Adaptor GGA3.
Traffic (Copenhagen, Denmark)
Ratcliffe CD,Sahgal P,Parachoniak CA,Ivaska J,Park M
Wed Jun 01 00:00:00 UTC 2016
SNX17 affects T cell activation by regulating TCR and integrin recycling.
Journal of immunology (Baltimore, Md. : 1950)
Osborne DG,Piotrowski JT,Dick CJ,Zhang JS,Billadeau DD
Fri May 01 00:00:00 UTC 2015
SNX17 affects T cell activation by regulating TCR and integrin recycling.
Journal of immunology (Baltimore, Md. : 1950)
Osborne DG,Piotrowski JT,Dick CJ,Zhang JS,Billadeau DD
Fri May 01 00:00:00 UTC 2015
SNX17 affects T cell activation by regulating TCR and integrin recycling.
Journal of immunology (Baltimore, Md. : 1950)
Osborne DG,Piotrowski JT,Dick CJ,Zhang JS,Billadeau DD
Fri May 01 00:00:00 UTC 2015
SNX17 affects T cell activation by regulating TCR and integrin recycling.
Journal of immunology (Baltimore, Md. : 1950)
Osborne DG,Piotrowski JT,Dick CJ,Zhang JS,Billadeau DD
Fri May 01 00:00:00 UTC 2015
LRP1 is a receptor for Clostridium perfringens TpeL toxin indicating a two-receptor model of clostridial glycosylating toxins.
Proceedings of the National Academy of Sciences of the United States of America
Schorch B,Song S,van Diemen FR,Bock HH,May P,Herz J,Brummelkamp TR,Papatheodorou P,Aktories K
Tue Apr 29 00:00:00 UTC 2014
LRP1 is a receptor for Clostridium perfringens TpeL toxin indicating a two-receptor model of clostridial glycosylating toxins.
Proceedings of the National Academy of Sciences of the United States of America
Schorch B,Song S,van Diemen FR,Bock HH,May P,Herz J,Brummelkamp TR,Papatheodorou P,Aktories K
Tue Apr 29 00:00:00 UTC 2014
Sorting nexin 31 binds multiple β integrin cytoplasmic domains and regulates β1 integrin surface levels and stability.
Journal of molecular biology
Tseng HY,Thorausch N,Ziegler T,Meves A,Fässler R,Böttcher RT
Tue Sep 09 00:00:00 UTC 2014
Sorting nexin 31 binds multiple β integrin cytoplasmic domains and regulates β1 integrin surface levels and stability.
Journal of molecular biology
Tseng HY,Thorausch N,Ziegler T,Meves A,Fässler R,Böttcher RT
Tue Sep 09 00:00:00 UTC 2014
Sorting nexin 31 binds multiple β integrin cytoplasmic domains and regulates β1 integrin surface levels and stability.
Journal of molecular biology
Tseng HY,Thorausch N,Ziegler T,Meves A,Fässler R,Böttcher RT
Tue Sep 09 00:00:00 UTC 2014
Sorting nexin 17 prevents lysosomal degradation of β1 integrins by binding to the β1-integrin tail.
Nature cell biology
Böttcher RT,Stremmel C,Meves A,Meyer H,Widmaier M,Tseng HY,Fässler R
Sun May 06 00:00:00 UTC 2012
Sorting nexin 17 prevents lysosomal degradation of β1 integrins by binding to the β1-integrin tail.
Nature cell biology
Böttcher RT,Stremmel C,Meves A,Meyer H,Widmaier M,Tseng HY,Fässler R
Sun May 06 00:00:00 UTC 2012
Adaptor protein sorting nexin 17 interacts with the scavenger receptor FEEL-1/stabilin-1 and modulates its expression on the cell surface.
Biochimica et biophysica acta
Adachi H,Tsujimoto M
Sat May 01 00:00:00 UTC 2010
Adaptor protein sorting nexin 17 interacts with the scavenger receptor FEEL-1/stabilin-1 and modulates its expression on the cell surface.
Biochimica et biophysica acta
Adachi H,Tsujimoto M
Sat May 01 00:00:00 UTC 2010