Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with mouse based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Alternative splice variants encoding different isoforms have been identified in this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ39019; neurite outgrowth multiadaptor RhoGAP protein; Rho GTPase-activating protein 33; Rho-type GTPase-activating protein 33; sorting nexin 26; Sorting nexin-26; Tc10/CDC42 GTPase-activating protein
Gene Aliases: ARHGAP33; NOMA-GAP; SNX26; TCGAP
UniProt ID: (Human) O14559
Entrez Gene ID: (Human) 115703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support