Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDMNSFSPMM PTSPLSMINQ IKFEDEPDLK DLFITVDEPE SHVTTIETFI TYRIITKTSR GEFDSSEFEV RRRYQDFLWL KGKLEEAHPT LIIPPLPEKF IVKGMVERFN DDFIETRRKA LHKFLNRIAD HPTLTFNEDF KIFLTAQAWE LSSHKKQGPG LLSRMGQTVR AVASSMRGVK NRPEEFMEMN NFIELFSQKI NLIDKISQRI YKEEREYFDE MKEYGPIHIL WSASEEDLVD TLKDVASCID RCCKATEKRM SGLSEALLPV VHEYVLYSEM LMGVMKRRDQ IQAELDSKVE VLTYKKADTD LCLATWESFL TSQTNLHLEE ASEDKP
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region like some family members, and its function is unknown. This gene results in two transcript variants encoding two distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sorting nexin-7
Gene Aliases: SNX7
UniProt ID: (Human) Q9UNH6
Entrez Gene ID: (Human) 51375
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support