Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene is a member of a small family of acyl coenzyme A:cholesterol acyltransferases. The gene encodes a membrane-bound enzyme localized in the endoplasmic reticulum that produces intracellular cholesterol esters from long-chain fatty acyl CoA and cholesterol. The cholesterol esters are then stored as cytoplasmic lipid droplets inside the cell. The enzyme is implicated in cholesterol absorption in the intestine and in the assembly and secretion of apolipoprotein B-containing lipoproteins such as very low density lipoprotein (VLDL). Several alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAT-2; acyl Co-A: cholesterol acyltransferase 2; acyl coenzyme A:cholesterol acyltransferase 2; acyl-CoA:cholesterol acyltransferase 2; Acyl-coenzyme A:cholesterol acyltransferase 2; Cholesterol acyltransferase 2; Sterol O-acyltransferase 2
Gene Aliases: ACACT2; ACAT2; ARGP2; SOAT2
UniProt ID: (Human) O75908
Entrez Gene ID: (Human) 8435
Molecular Function:
acyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support