Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: DLVELKSLQ QQLSDVCYRQ ASQLEFRQNL LQAALEFHGV AQDLSQQLDG LLGMLCVDVA PADGASIQQT LKLLEEKLKS VDVGLQGLRE KGQGLLDQIS NQASWAYGKD VTIENKENVD HIQGVMEDMQ LRKQRCEDMV DVRRLKMLQM VQLFKCEEDA AQAVEWLSEL LDALLKTHIR LGDDAQETKV LLEKHRKFVD VAQSTYDYGR QLLQATVVLC QSLRCTSRSS GDTLPRLNRV WKQFTIASEE RVHRLEMAIA FHSNAEKILQ DCPEEPEAIN DEEQFDEIEA VGKSLLDRLT VPVVYPDGTE QYFGSPSDMA STAENIRDRM KLVNLKRQQL RHPEMVTTES (348-696 aa encoded by BC047578)
SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Huntingtin-interacting protein-like protein; Protein Solo; SEC14 and spectrin domains 1; SEC14 domain and spectrin repeat-containing protein 1
Gene Aliases: 1500031J16Rik; A930010O20; RGD1562244; SESTD1; SOLO; Solo/Trio8
UniProt ID: (Human) Q86VW0, (Mouse) Q80UK0
Entrez Gene ID: (Human) 91404, (Mouse) 228071, (Rat) 295678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support