Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
Son of sevenless (SOS) is a guanine nucleotide exchange factor that activates Ras in response to growth factor stimulation. Sos1, and its closely related Sos2 paralog, are comprised of an N-terminal histone-binding domain, a RacGEF catalytic (DH) domain juxtaposed with a PH domain, a Ras exchanger motif (REM) domain, a RasGEF catalytic domain, and a C-terminal proline-rich region. Sos1 catalyzes the GDP-GTP exchange within the membrane-bound GTPase Ras and thereby switches on a key signaling circuit that involves the activation of MAPK cascade central to cellular proliferation, survival and differentiation. Growth factor stimulation rapidly induces the phosphorylation of Sos on multiple serine and threonine sites. Phosphorylation of residues within the Sos C-terminus by the MAP/ERK kinase disrupts the Grb2-Sos complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G GINGF; gingival fibromatosis; gingival fibromatosis, hereditary, 1; guanine nucleotide exchange factor; Son of sevenless homolog 1; SOS-1
Gene Aliases: GF1; GGF1; GINGF; HGF; NS4; SOS1
UniProt ID: (Human) Q07889
Entrez Gene ID: (Human) 6654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support