Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALPGSSQD QAWSLEPPAA TAAASSSSGP QEREGAGSPA APGTLPLEKV KRPMNAFMVW SSAQRRQMAQ QNPKMHNSEI SKRLGAQWKL LDEDEKRPFV EEAKRLRARH LRDYPDYKYR PRRKAKSSGA GPSRCGQGRG NLASGGPLWG PGYATTQPSR GFGYRPPSYS TAYLPGSYGS SHCKLEAPSP CSLPQSDPRL QGELLPTYTH YLPPGSPTPY NPPLAGAPMP LTHL (1-233 aa encoded by BC000985)
SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SOX-12; protein SOX-15; Protein SOX-20; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15; SRY-box containing gene 15; SRY-box transcription factor 15; Transcription factor SOX-15
Gene Aliases: Sox-15; SOX12; SOX15; Sox16; SOX20; SOX26; SOX27
UniProt ID: (Human) B4DWU7, (Mouse) P43267
Entrez Gene ID: (Human) 6665, (Mouse) 20670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support