Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SOX-12; Protein SOX-15; Protein SOX-20; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15
Gene Aliases: SOX12; SOX15; SOX20; SOX26; SOX27
UniProt ID: (Human) O60248
Entrez Gene ID: (Human) 6665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support