Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MRQRLLPSVT SLLLVALLFP GSSQARHVNH SATEALGELR ERAPGQGTNG
Sequence homology: Human: 100%
SPAG11B is one of several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides.This gene encodes several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides. The gene is located on chromosome 8p23 near the defensin gene cluster. Alternative splicing of this gene results in seven transcript variants encoding different isoforms. Two different N-terminal and five different C-terminal protein sequences are encoded by the splice variants. Two additional variants have been described, but their full length sequences have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antimicrobial-like protein Bin-1b; epididymal protein 2B; He2; Human epididymis-specific protein 2; MGC61846; Protein EP2; Sperm antigen HE2; Sperm-associated antigen 11A; Sperm-associated antigen 11B
Gene Aliases: BIN1B; EDDM2B; EP2; EP2C; EP2D; HE2; HE2C; SPAG11; SPAG11A; SPAG11B
UniProt ID: (Human) Q6PDA7
Entrez Gene ID: (Human) 10407
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support