Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene maps to chromosome X in a head-to-tail orientation with SPANX family member B2, which appears to be a duplication of the B1 locus. The SPANXB genes are unique members of this gene family, since they contain an additional 18 nt in their coding region compared to the majority of family members. Although the protein encoded by this gene contains consensus nuclear localization signals, the major site for subcellular localization of expressed protein is in the cytoplasmic droplets of ejaculated spermatozoa. This protein provides a biochemical marker for studying the unique structures in spermatazoa, while attempting to further define its role in spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 11.2; cancer/testis antigen family 11, member 2; CT11.2; Nuclear-associated protein SPAN-Xb; nuclear-associated protein SPAN-Xf; SPANX family member B/F; SPANX family member B1; SPANX family member F1; SPANX family, member B1; SPANX family, member B2; SPANX family, member F1; SPANX family, member F2; SPANX-B; sperm protein associated with the nucleus on the X chromosome B/F; Sperm protein associated with the nucleus on the X chromosome B1; sperm protein associated with the nucleus, X chromosome, family member B1
Gene Aliases: B1; CT11.2; SPANX-B; SPANXB; SPANXB1; SPANXB2; SPANXF1; SPANXF2
UniProt ID: (Human) Q9NS25
Entrez Gene ID: (Human) 728695
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support