Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MGSASPGLSS VSPSHLLLPP DTVSRTGLEK AAAGAVGLER RDWSPSPPAT PEQGLSAFYL SYFDMLYPED SSWAAKAPGA SSREEPPEEP EQCPVIDSQA PAGSLDLVPG GLTLEEHSLE QVQSMVVGEV LKDIETACKL LNITADPMDW SPSNVQKWLL WTEHQYRLPP MGKAFQELAG KELCAMSEEQ FRQRSPLGGD VLHAHLDIWK SAAWMKERTS PGAIHYCAST SEESWTDSEV DSSCSGQPIH LWQFLKELLL KPHSYGRFIR WLNKEKGIFK IEDSAQVARL WGIRKNRPAM NYDKLSRSIR QYYKKGIIRK PDISQRLVYQ FVHPI
PDEF (prostate-derived Ets factor) is a 37.5-kDa protein that acts as an androgen-independent transcriptional activator for the PSA (prostate specific antigen) promoter. It also interacts directly with the DNA binding domain of androgen receptor and enhances androgen-mediated activation of the PSA promoter. PDEF was shown to have transcript levels 192-fold higher in the peripheral blood of some breast cancer patients in comparison with normal individuals. PDEF mRNA is also frequently over-expressed in human breast tumors as well as in human ovarian tumors. In ovarian tumors, PDEF expression seems to down-modulate malignant potential, and thus provides a rationale to screen for drugs that induce PDEF expression in epithelial ovarian tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Prostate epithelium-specific Ets transcription factor; Prostate-derived Ets factor; Prostate-specific Ets; SAM pointed domain-containing Ets transcription factor
Gene Aliases: bA375E1.3; PDEF; PSE; SPDEF
UniProt ID: (Human) O95238
Entrez Gene ID: (Human) 25803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support