Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a mitochondrial metalloprotease protein that is a member of the AAA family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Mutations in this gene cause autosomal recessive spastic paraplegia 7. Two transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell adhesion regulator; Cell matrix adhesion regulator; Paraplegin; paraplegin, isoform 1; spastic paraplegia 7 (pure and complicated autosomal recessive); Spastic paraplegia 7 protein; SPG7, paraplegin matrix AAA peptidase subunit
Gene Aliases: AI452278; AU015315; CAR; CMAR; PGN; SPG5C; SPG7
UniProt ID: (Human) Q9UQ90, (Mouse) Q3ULF4, (Rat) Q7TT47
Entrez Gene ID: (Human) 6687, (Dog) 489657, (Mouse) 234847, (Rat) 353231
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support