Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: MDPAGGPRGV LPRPCRVLVL LNPRGGKGKA LQLFRSHVQP LLAEAEISFT LMLTERRNHA RELVRSEELG RWDALVVMSG DGLMHEVVNG LMERPDWETA IQKPLCSLPA GSGNALAASL NHYAGYEQVT NEDLLTNCTL LLCRRLLSPM NLLSLHTASG LRLFSVLSLA WGFIADVDLE SEKYRRLGEM RFTLGTFLLL AALRTYRGRL AYLPVGRVGS KTPASPVVVQ QGPVDAHLVP LEEPVPSHWT VVPDEDFVLV LALLHSHLGS EMFAAPMGRC AAGVMHLFYV RAGVSRAMLL RLFLAMEKGR HMEYECPYLV YVPVVAFRLE PKDGKGVFAV DGELMVSEAV QGQVHPNYFW MVSGCVEPPP SWKPQQMPPP EEPL
SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signaling molecule sphingosine-1-phosphate (S1P). S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapeutics for the treatment of cancer. SPHK1 catalyzes the phosphorylation of sphingosine to sphingosine 1 phosphate (S1P), an important signaling molecule with intra- and extracellular functions. Inside the cell S1P acts as a signaling molecule like other sphingolipid metabolites like ceramide and sphingosine. S1P has been implicated in regulating cell differentiation, calcium mobilization from intracellular stores, and apoptosis. The cell surface receptors for S1P are the EDG family of G protein-coupled receptors (S1P Receptors).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetyltransferase SPHK1; RP23-193A16.4; SK 1; SKase; Sphingosine kinase 1; SPK 1
Gene Aliases: SK1; SPHK; SPHK1; SPK
UniProt ID: (Human) Q9NYA1
Entrez Gene ID: (Human) 8877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support