Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: U-87MG, A431, HeLa, mouse liver, mouse brain, mouse testis, rat brain, rat testis. The target is usually found in the following locations: Cell membrane, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle membrane, Peripheral membrane protein, cytoskeleton, cytosol, perinuclear region.
Immunogen sequence: MARAGSCGGA AAGAGRPEPW ELSLEEVLKA YEQPLNEEQA WAVCFQGCRG LRGSPGRRLR DTGDLLLRGD GSVGAREPEA AEPATMVVPL ASSEAQTVQS LGFAIYRALD WGLDESEERE LSPQLERLID LMANNDSEDS GCGAADEGYG GPEEEEEAEG VPRSVRTFAQ AMRLCAARLT DPRGAQAHYQ
Acts as an actin nucleation factor, remains associated with the slow-growing pointed end of the new filament (PubMed:21620703). Involved in intracellular vesicle transport along actin fibers, providing a novel link between actin cytoskeleton dynamics and intracellular transport. Required for asymmetric spindle positioning and asymmetric cell division during meiosis (PubMed:21620703). Required for normal formation of the cleavage furrow and for polar body extrusion during female germ cell meiosis (PubMed:21620703). Also acts in the nucleus: together with SPIRE1 and SPIRE2, promotes assembly of nuclear actin filaments in response to DNA damage in order to facilitate movement of chromatin and repair factors after DNA damage (PubMed:26287480). [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein spire homolog 2; Spir-2; spire actin nucleation factor 2; spire family actin nucleation factor 2; spire homolog 2; spire-type actin nucleation factor 2
Gene Aliases: BC026502; KIAA1832; Spir-2; SPIR2; SPIRE2
UniProt ID: (Human) Q8WWL2, (Mouse) Q8K1S6
Entrez Gene ID: (Human) 84501, (Rat) 307925, (Mouse) 234857
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support