Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
SPOCK2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ97039; KIAA0275CWCV; SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 2; Testican-2
Gene Aliases: KIAA0275; SPOCK2; testican-2; TICN2; UNQ269/PRO306
UniProt ID: (Human) Q92563
Entrez Gene ID: (Human) 9806
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support