Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLKVSAVLCV CAAAWCSQSL AAAAAVAAAG GRSDGGNFLD DKQWLTTISQ YDKEVGQWNK FRDDDYFRTW SPGKPFDQAL DPAKDPCLKM KCSRHKVCIA QDSQTAVCIS HRRLTHRMKE AGVDHRQWRG PILSTCKQCP VVYPSPVCGS DGHTYSFQCK LEYQACVLGK QISVKCEGHC PCPSDKPTST SRNVKRACSD LEFREVANRL RDWFKALHES GSQNKKTKTL LRPERSRFDT SILPICKDSL GWMFNRLDTN YDLLLDQSEL RSIYLDKNEQ CTKAFFNSCD TYKDSLISNN EWCYCFQRQQ DPPCQTELSN IQKRQGVKKL LGQYIPLCDE DGYYKPTQCH GSVGQCWCVD RYGNEVMGSR INGVADCAID FEISGDFASG DFHEWTDDED DEDDIMNDED EIEDDDEDEG DDDDGGDDHD VYI
Proteoglycans, which consist of a core protein and covalently linked glycosaminoglycans, are components of the extracellular matrix. SPOCK3 encodes a member of a novel Ca(2+)-binding proteoglycan family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 3; Testican-3
Gene Aliases: HSAJ1454; SPOCK3; TES-3; TICN3; UNQ409/PRO771
UniProt ID: (Human) Q9BQ16
Entrez Gene ID: (Human) 50859
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support