Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain.
Predicted reactivity: Mouse (90%), Rat (89%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Proteoglycans, which consist of a core protein and covalently linked glycosaminoglycans, are components of the extracellular matrix. SPOCK3 encodes a member of a novel Ca(2+)-binding proteoglycan family.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 3; testican 3; Testican-3
Gene Aliases: 2900045C01Rik; AI428471; HSAJ1454; mKIAA4039; SPOCK3; TES-3; TICN3; UNQ409/PRO771
UniProt ID: (Human) Q9BQ16, (Mouse) Q8BKV0
Entrez Gene ID: (Human) 50859, (Mouse) 72902
Molecular Function: extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support