Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RGPILSTCKQ CPVVYPSPVC GSDGHTYSFQ CKLEYQACVL GKQISVKCEG HCPCPSDKPT STSRNVKRAC SDLEFREVAN RLRDWFKALH ESGSQNKKT
Proteoglycans, which consist of a core protein and covalently linked glycosaminoglycans, are components of the extracellular matrix. SPOCK3 encodes a member of a novel Ca(2+)-binding proteoglycan family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SPARC/osteonectin, CWCV, and Kazal-like domains proteoglycan 3; Testican-3
Gene Aliases: HSAJ1454; SPOCK3; TES-3; TICN3; UNQ409/PRO771
UniProt ID: (Human) Q9BQ16
Entrez Gene ID: (Human) 50859
Molecular Function:
extracellular matrix glycoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support