Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Catalyzes intramembrane proteolysis of some signal peptides after they have been cleaved from a preprotein, resulting in the release of the fragment from the ER membrane into the cytoplasm. Required to generate lymphocyte cell surface (HLA-E) epitopes derived from MHC class I signal peptides. May play a role in graft rejection. May be necessary for the removal of the signal peptide that remains attached to the hepatitis C virus core protein after the initial proteolytic processing of the polyprotein. Involved in the intramembrane cleavage of the integral membrane protein PSEN1. Cleaves the integral membrane protein XBP1 isoform 1 in a DERL1/RNF139- dependent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hIMP1; histocompatibility (minor) 13; histocompatibility 13; IMP-1; Intramembrane protease 1; minor histocompatibility antigen 13; Minor histocompatibility antigen H13; Presenilin-like protein 3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1
Gene Aliases: H13; HM13; IMP1; IMPAS; IMPAS-1; MSTP086; PSENL3; PSL3; SPP; SPPL1
UniProt ID: (Human) Q8TCT9
Entrez Gene ID: (Human) 81502
Molecular Function: aspartic protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support