Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Human: 100%
Intramembrane-cleaving aspartic protease (I-CLiP) that may be able to cleave type II membrane signal peptides in the hydrophobic plane of the membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IMP-5; Intramembrane protease 5; Signal peptide peptidase-like 2C; SPP-like 2C
Gene Aliases: IMP5; SPPL2C
UniProt ID: (Human) Q8IUH8
Entrez Gene ID: (Human) 162540
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support