Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
SPRN has PrP(C)-like neuroprotective activity. SPRN may act as a modulator for the biological actions of normal and abnormal PrP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hypothetical protein BC004409; Protein shadoo; Shadow of prion protein
Gene Aliases: bA108K14.1; SHADOO; SHO; SPRN
UniProt ID: (Human) Q5BIV9
Entrez Gene ID: (Human) 503542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support