Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total SPZ1.
Spermatogenic leucine zipper protein 1 is a member of the basic helix-loop-helix (bHLH) family of transcription factors which is expressed specifically in the testis and epididymis. Studies show that SPZ1 acts downstream of mitogen-activated protein kinase (MAPK), an extracellular signal-regulated kinase 1/2 to up-regulate cell proliferation and tumorigenesis. Also, through an interaction with proliferating cell nuclear antigen (PCNA) promoter, SPZ1 induced cell proliferation concomitant with an increase in PCNA gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 1, regulatory subunit 148; Spermatogenic leucine zipper protein 1; spermatogenic Zip 1; testis secretory sperm-binding protein Li 232m; Testis-specific protein 1; Testis-specific protein NYD-TSP1
Gene Aliases: NYD-TSP1; PPP1R148; SPZ1; TSP1
UniProt ID: (Human) Q9BXG8, (Rat) Q6AXY9
Entrez Gene ID: (Human) 84654, (Rat) 499510
Molecular Function:
basic leucine zipper transcription factor
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support