Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Suggested positive control: mouse liver lysate or protein.
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; CD36-related class B scavenger receptor haSR-BI; CLA-1; Collagen type I receptor, thrombospondin receptor-like 1; HaSR-BI; HDL QTL 1; high density lipoprotein receptor; scavanger receptor class B type 1; scavenger receptor b1; Scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SR-BI; SRB1
Gene Aliases: AI120173; CD36; CD36L1; CLA-1; CLA1; D5Ertd460e; haSR-bI; Hdlq1; HDLQTL6; Hlb398; mSR-BI; SCARB1; SR-B1; SR-BI; SRB1; SRBI
UniProt ID: (Bovine) O18824, (Human) Q8WTV0, (Rat) P97943, (Mouse) Q61009, (Chinese hamster) Q60417
Entrez Gene ID: (Bovine) 282346, (Human) 949, (Rat) 25073, (Mouse) 20778, (Chinese hamster) 100736552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support