Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: HeLa, 293T, SGC-7901.
Immunogen sequence: VARSSTLPPS NSNMVSTRIN RQQSSDLHSS SHSNSSNSQG SFGCSPGSQI VANVALNQGQ ASSQSSNPSL NLNNSPMEGT GISLAQFMSP RRQVTSGLAT RPRMPNNSFP PNISTLSSPV GMTSSACNNN NRSYSNIPVT SLQGMNEGPN NSVGFSASSP VLRQMSSQNS PSRLNIQPAK AESKDNKEIA SILNEMIQSD NSSSDGKPLD SGLLHNNDRL SDGDSKYSQT SHKLVQLLTT TAEQQLRHAD IDTSCKDVLS CTGTSNSASA NSSGGSCPSS HSSLTERHKI LHRLLQEGSP SDITTLSVEP DKKDSASTSV SVTGQVQGNS SIKLELDASK KKESKDHQLL R
Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASV; bHLHe74; c-SRC; Class E basic helix-loop-helix protein 74; Hin-2 protein; NCoA-1; Nuclear receptor coactivator 1; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; SRC-1; SRC-1 (-Q); Steroid receptor coactivator 1; steroid receptor coactivator-1
Gene Aliases: bHLHe42; BHLHE74; F-SRC-1; KAT13A; NCOA1; RIP160; SRC1
UniProt ID: (Human) Q15788
Entrez Gene ID: (Human) 8648
Molecular Function: histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support