Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 7
PA1-846 detects glucocorticoid receptor interacting protein-1 (GRIP1) from hamster and human cells.
PA1-846 has been successfully used in immunocytochemistry, and immunofluorescence procedures. Immunocytochemical staining of GRIP1 in CHO cells with PA1-846 results in cytoplasmic and nuclear staining. This antibody has not been used successfully in Western blot procedures.
PA1-846 immunogen is a synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1. The PA1-846 immunizing peptide (Cat. # PEP-050) is available for use in neutralization and control experiments.
SRC2 (steroid receptor coactivator 2), also known as nuclear receptor coactivator2, NCOA2 or TIF2, aids in the function of nuclear hormone receptors. Nuclear hormone receptors are conditional transcription factors that play important roles in various aspects of cell growth, development, and homeostasis by controlling expression of specific genes. Members of the nuclear hormone receptor superfamily, which includes the 5 steroid receptors and class II nuclear receptors, are structurally characterized by 3 distinct domains: an N-terminal transcriptional activation domain, a central DNA-binding domain, and a C-terminal hormone-binding domain. Before the binding of hormone, steroid receptors, which are sometimes called class I of the nuclear hormone receptor family, remain inactive in a complex with heat-shock protein-90 and other stress family proteins. Binding of hormone induces critical conformational changes in steroid receptors that cause them to dissociate from the inhibitory complex, bind as homodimers to specific DNA enhancer elements associated with target genes, and modulate that gene's transcription. After binding to enhancer elements, transcription factors require transcriptional coactivator proteins to mediate their stimulation of transcription initiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHe75; c-fgr; c-src2; Class E basic helix-loop-helix protein 75; FLJ43153; glucocorticoid receptor-interacting protein-1; hTIF2; MGC75096; NCoA-2; Nuclear receptor coactivator 2; p160 steroid receptor coactivator 2; p55c-fgr; p58c-fgr; TIF 2; Transcriptional intermediary factor 2
Gene Aliases: BHLHE75; GRIP1; I79_019839; KAT13C; NCoA-2; NCOA2; SRC2; TIF2
UniProt ID: (Human) Q15596
Entrez Gene ID: (Human) 10499, (Chinese hamster) 100762955
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support