Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MMQKLLKCSR LVLALALILV LESSVQGYPT QRARYQWVRC NPDSNSANCL EEKGPMFELL PGESNKIPRL RTDLFPKTRI QDLNRIFPLS EDYSGSGFGS GSGSGSGSGS GFLTEMEQDY QLVDESDAFH DNLRSLDRNL PSDSQDLGQH GLEEDFML
This gene encodes a protein best known as a hematopoietic cell granule proteoglycan. Proteoglycans stored in the secretory granules of many hematopoietic cells also contain a protease-resistant peptide core, which may be important for neutralizing hydrolytic enzymes. This encoded protein was found to be associated with the macromolecular complex of granzymes and perforin, which may serve as a mediator of granule-mediated apoptosis. Two transcript variants, only one of them protein-coding, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; Hematopoietic proteoglycan core protein; P.PG; Platelet proteoglycan core protein; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; secretory granule proteoglycan core peptide; Secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan
Gene Aliases: PPG; PRG; PRG1; SRGN
UniProt ID: (Human) P10124
Entrez Gene ID: (Human) 5552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support