Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: EVFTYGQCP YEGMTNHETL QQIMRGYRLP RPAACPAEVY VLMLECWRSS PEERPSFATL REKLHAIHRC HP (418-488 aa encoded by BC153198)
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tyrosine-protein kinase Srms
Gene Aliases: C20orf148; dJ697K14.1; PTK70; SRM; SRMS
UniProt ID: (Human) Q9H3Y6
Entrez Gene ID: (Human) 6725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support