Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Control: HepG2, rat brain tissue, human colon cancer tissue, human kidney tissue, mouse small intestine tissue.
Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Signal recognition particle 54 kDa protein; signal recognition particle 54kD; signal recognition particle 54kDa; Signal recognition particle subunit SRP54
Gene Aliases: SRP54
UniProt ID: (Human) P61011
Entrez Gene ID: (Human) 6729
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support