Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPQYQTWEE FSRAAEKLYL ADPMKARVVL KYRHSDGNLC VKVTDDLVCL VYKTDQAQDV KKIEKFHSQL MRLMVAKEAR NVTMETE (1-86 aa encoded by B C015094)
Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Signal recognition particle 9 kDa protein; signal recognition particle 9kD; signal recognition particle 9kDa; SRP9
Gene Aliases: 9kDa; ALURBP; SRP9
UniProt ID: (Human) P49458, (Mouse) P49962
Entrez Gene ID: (Human) 6726, (Rat) 690345, (Mouse) 27058
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support