Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MERKVLALQA RKKRTKAKKD KAQRKSETQH RGSAPHSESD LPEQEEEILG SDDDEQEDPN DYCKGGYHLV KIGDLFNGRY HVIRKLGWGH FSTVWLSWDI QGKKFVAMKV VKSAEHYTET ALDEIRLLKS VRNSDPNDPN REMVVQLLDD FKISGVNGTH ICMVFEVLGH HLLKWIIKSN YQGLPLPCVK KIIQQVLQGL DYLHTKCRIT HTDIKPENIL LSVNEQYIRR LAAEATEWQR SGAPPPSGSA VSTAPQPKPA DKMSKNKKKK LKKKQKRQAE LLEKRMQEIE EMEKESGPGQ KRPNKQEESE SPVERPLKEN PPNKMTQEKL EESSTIGQDQ TLMERDTEGG AAEINCNGVI EVINYTQNSN NETLRHKEDL HNANDCDVQN LNQESSFLSS QNGDSSTSQE TDSCTPITSE VSDTMVCQSS STVGQSFSEQ HISQLQESIR AEIPCEDEQE QEHNGPLDNK GKSTAGNFLV NPLEPKNAEK LKVKIADLGN ACWVHKHFTE DIQTRQYRSL EVLIGSGYNT PADIWSTACM AFELATGDYL FEPHSGEEYT RDEDHIALII ELLGKVPRKL IVAGKYSKEF FTKKGDLKHI TKLKPWGLFE VLVEKYEWSQ EEAAGFTDFL LPMLELIPEK RATAAECLRH PWLNS
SRPK1 is a serine/threonine kinase that plays a role in the regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. SRPK1 localizes to the nucleus and the cytoplasm. It is thought to play a role in regulation of both constitutive and alternative splicing by regulating intracellular localization of splicing factors. A second alternatively spliced transcript variant for this gene has been described, but its full length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000016277; Serine/arginine-rich protein-specific kinase 1; serine/arginine-rich splicing factor kinase 1; serine/threonine-protein kinase SRPK1; SFRS protein kinase 1; SR-protein-specific kinase 1; SRPK1; SRSF protein kinase 1
Gene Aliases: SFRSK1; SRPK1
UniProt ID: (Human) Q96SB4
Entrez Gene ID: (Human) 6732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support