Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
The gene encodes a subunit of the endoplasmic reticulum signal recognition particle receptor that, in conjunction with the signal recognition particle, is involved in the targeting and translocation of signal sequence tagged secretory and membrane proteins across the endoplasmic reticulum. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Docking protein alpha; DP-alpha; MGC17355; MGC3650; MGC9571; signal recognition particle receptor (docking protein); Signal recognition particle receptor subunit alpha; SR-alpha; SRP-alpha
Gene Aliases: 1300011P19Rik; D11Mgi27; DP; Sralpha; SRPR; SRPRA
UniProt ID: (Human) P08240, (Mouse) Q9DBG7
Entrez Gene ID: (Human) 6734, (Mouse) 67398
Molecular Function:
G-protein
protein-binding activity modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support