Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Antibody detects endogenous levels of total SRR.
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-serine ammonia-lyase; D-serine dehydratase; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase
Gene Aliases: ILV1; ISO1; SRR; Srs
UniProt ID: (Human) Q9GZT4, (Rat) Q76EQ0, (Mouse) Q9QZX7
Entrez Gene ID: (Human) 63826, (Rat) 303306, (Mouse) 27364
Molecular Function:
epimerase/racemase
isomerase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support