Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSPEVDSKSR LSPRRSRSGS SPEVKDKPRA APRAQSGSDS SPEPKAPAPR ALPRRSRSGS SSKGRGPSPE GSSSTESSPE HPPKSRTARR GSRSSPEPKT KSRTPPRRRS SRSSPELTRK ARLSRRSRSA SSSPETRSRT PPRHRRSPSV SSPEPAEKSR SSRRRRSASS PRTKTTSRRG RSPSPKPRGL QRSRSRSRRE KTRTTRRRDR SGSSQ
SRRM2 is involved in pre-mRNA splicing. It may function at or prior to the first catalytic step of splicing at the catalytic center of the spliceosome and may do so by stabilizing the catalytic center or the position of the RNA substrate. It binds to RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 300 kDa nuclear matrix antigen; RNA binding protein; ser/Arg-related nuclear matrix protein of 300 kDa; Serine/arginine repetitive matrix protein 2; Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa; Splicing coactivator subunit SRm300; SR-related nuclear matrix protein of 300 kDa; Tax-responsive enhancer element-binding protein 803; TaxREB803; testicular secretory protein Li 53
Gene Aliases: 300-KD; Cwc21; CWF21; HSPC075; KIAA0324; SRL300; SRM300; SRRM2
UniProt ID: (Human) Q9UQ35
Entrez Gene ID: (Human) 23524
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support