Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/arginine repetitive matrix protein 3; SRRM2-like protein
Gene Aliases: 2900083I11Rik; AB041661; MNCb-4760; RGD1307391; SRm300-like; Srrm2l; Srrm3
UniProt ID: (Mouse) Q9JJ91
Entrez Gene ID: (Human) 222183, (Rat) 685890, (Mouse) 58212
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support