Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSGWADERGG EGDGRIYVGN LPTDVREKDL EDLFYKYGRI REIELKNRHG LVPFAFVRFE DPRDAEDAIY GRNGYDYGQC RLRVEFPRTY GGRGGWPRGG RNGPPTRRSD FRVLVSGLPP SGSWQDLKDH MREAGDVCYA DVQKDGVGMV EYLRKEDMEY ALRKLDDTKF RSHEGETSYI RVYPERSTSY GYSRSRSGSR GRDSPYQSRG SPHYFSPFRP Y; Positive Samples: BT-474, HT-29, HepG2, Mouse spleen, Mouse testis; Cellular Location: Nucleus
The protein encoded by this gene is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two pseudogenes, one on chromosome 15 and the other on chromosome 21, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pre-mRNA-splicing factor SRp30C; Serine/arginine-rich splicing factor 9; Splicing factor, arginine/serine-rich 9; SR splicing factor 9
Gene Aliases: 25kDa; 2610029M16Rik; SFRS9; SRP30C; SRSF9
UniProt ID: (Human) Q13242, (Mouse) Q9D0B0
Entrez Gene ID: (Human) 8683, (Mouse) 108014
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support