Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MFAKGKGSAV PSDGQAREKL ALYVYEYLLH VGAQKSAQTF LSEIRWEKNI TLGEPPGFLH SWWCVFWDLY CAAPERRDTC EHSSEAKAFH DYSAAAAPSP VL
May be involved in transcription regulation of the alpha 2(I) collagen gene where it binds to the single-stranded polypyrimidine sequences in the promoter region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sequence-specific single-stranded-DNA-binding protein; Single-stranded DNA-binding protein 3
Gene Aliases: CSDP; SSBP3; SSDP; SSDP1
UniProt ID: (Human) Q9BWW4
Entrez Gene ID: (Human) 23648
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support