Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: TYLVAFAYKN VKFVLKHKVA QKREDAVSKE VTRKLSEADN RKMSRKEKDE RILWKKNEVA DYEATTFSIF Y
SSR3 (signal sequence receptor, gamma) is also known as TRAPG. By immunoaffinity chromatography, biochemical purification, peptide sequence analysis, and screening of a rat liver cDNA library, Hartmann et al. (1993) obtained a cDNA encoding Ssr3, which they termed Trap-gamma. The deduced 185-amino acid protein spans the membrane 4 times and is expressed as a 20-kD protein. Gross (2012)mapped the SSR3 gene to chromosome 3q25.31 based on an alignment of the SSR3 sequence with the genomic sequence (GRCh37). Hartmann et al. (1993)found that Ssr3 was associated in equal amounts with Ssr1, Ssr2, and Ssr4 in canine pancreatic microsomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Signal sequence receptor subunit gamma; signal sequence receptor, gamma (translocon-associated protein gamma); SSR gamma; SSR-gamma; translocon-associated protein gamma; translocon-associated protein gamma subunit; Translocon-associated protein subunit gamma; TRAP-complex gamma subunit; TRAP-gamma
Gene Aliases: 0610038P07Rik; AL022999; AU022074; AW553833; SSR3; TRAPG
UniProt ID: (Human) Q9UNL2, (Mouse) Q9DCF9, (Rat) Q08013
Entrez Gene ID: (Human) 6747, (Mouse) 67437, (Rat) 81784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support