Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Control: 293T, A431, HeLa, HepG2
Predicted Reactivity: Mouse (99%), Zebrafish (90%), Xenopus laevis (93%), Dog (98%), Chicken (98%), Rhesus Monkey (100%), Bovine (99%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SSU72 is a highly conserved homologue of yeast Ssu72, a CTD phosphatase and a component of the polyadenylation/ termination machinery. The human Ssu72 is located on Chromosome 1p36 and seems to represent a single gene. It was identified as a pRb binding factor and was also shown to interact with TFIIB and yeast PTA1. The ABs recognize a single band of approximately 30 Kda on immunoblots of various human cell lines but do not work well on mouse tissues.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTD phosphatase SSU72; FLJ13048; RNA polymerase II subunit A C-terminal domain phosphatase SSU72; RP5-832C2.2
Gene Aliases: 1190002E22Rik; 1500011L16Rik; 2610101M12Rik; HSPC182; PNAS-120; RGD1307854; SSU72
UniProt ID: (Human) Q9NP77, (Rat) Q4KLK9, (Mouse) Q9CY97
Entrez Gene ID: (Human) 29101, (Rat) 298681, (Mouse) 68991
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support