Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Repressor that binds to DNA sequences containing a bipartite element consisting of a direct repeat of the sequence 5'-AAAGTTT-3' separated by 2-9 nucleotides. Represses basal transcription activity from target promoters. Inhibits colony formation in cultured breast cancer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2-HC type zinc finger protein r-MyT3; Neural zinc finger factor 3; neural zinc finger protein NZF-3; neural zinc finger transcription factor 3; NZF-3; ST18, C2H2C-type zinc finger; suppression of tumorigenicity 18 (breast carcinoma) (zinc finger protein); Suppression of tumorigenicity 18 protein; suppression of tumorigenicity 18, zinc finger; suppression of tumorigenicity protein 18; Zinc finger protein 387
Gene Aliases: AV348974; KIAA0535; mKIAA0535; Myt3; NZF-3; NZF3; r-MyT3; ST18; ZC2H2C3; ZC2HC10; ZNF387
UniProt ID: (Human) O60284, (Mouse) Q80TY4, (Rat) Q9QX27
Entrez Gene ID: (Rabbit) 100351142, (Human) 9705, (Mouse) 240690, (Rat) 266680
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support