Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Recommended positive controls: NT2D1, IMR32.
Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (90%), Pig (100%), Chicken (89%), Chimpanzee (100%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi apparatus but can be proteolytically processed to a soluble form. This protein is a member of glycosyltransferase family 29. Multiple transcript variants encoding several different isoforms have been found for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 2,3-sialyltransferase III; Alpha 2,3-ST 3; alpha-2,3-sialyltransferase II; Beta-galactoside alpha-2,3-sialyltransferase 3; CMP-N-acetylneuraminate-beta-1,4-galactoside alpha-2,3-sialyltransferase; Gal beta-1,3(4) GlcNAc alpha-2,3 sialyltransferase; Gal beta-1,3(4)GlcNAc alpha-2,3 sialyltransferase; mental retardation, non-syndromic, autosomal recessive, 12; N-acetyllactosaminide alpha-2,3-sialyltransferase; Sialyltransferase 6; sialyltransferase 6 (N-acetyllacosaminide alpha 2,3-sialyltransferase); ST3Gal III; ST3GalIII; ST3N
Gene Aliases: EIEE15; MRT12; SIAT6; ST3GAL3; ST3GALII; ST3GalIII; ST3N
UniProt ID: (Human) Q11203
Entrez Gene ID: (Human) 6487
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support