Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is Hela cell lysate.
PA5-20748 can be used with blocking peptide PEP-0862.
Sialyltransferases catalyze the transfer of sialic acid from cytidine 5-prime monophospho-N-acetylneuraminic acid (CMP-NeuAc) to terminal positions of glycoprotein and glycolipid carbohydrate groups. Terminal NeuAc residues are key determinants of carbohydrate structures, such as the sialyl-Lewis X determinants, and are widely distributed in many cell types. However, cancer cells often express more heavily sialylated glycans on their cell surface and this feature sometimes correlates with invasiveness. In contrast, expression of ST3gal6, a member of the sialyltransferase family that sialylates type II lactosamine structures on glycoproteins and glycolipids, was found to be significantly decreased by hypermethylation of the gene in gastrointestinal cancer. At least three isoforms of ST3gal6 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-2,3-sialyltransferase VI; alpha2,3-sialyltransferase ST3Gal VI; CMP-NeuAc:beta-galactoside alpha-2,3-sialyltransferase VI; sia10; Sialyltransferase 10; sialyltransferase 10 (alpha-2,3-sialyltransferase VI); ST3Gal VI; ST3GalVI; Type 2 lactosamine alpha-2,3-sialyltransferase
Gene Aliases: 1700023B24Rik; AI930218; AW552396; SIAT10; ST3GAL6; ST3GALVI
UniProt ID: (Human) Q9Y274, (Mouse) Q8VIB3
Entrez Gene ID: (Human) 10402, (Rat) 304023, (Mouse) 54613
Molecular Function:
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support