Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Transfers sialic acid from the donor of substrate CMP-sialic acid to galactose containing acceptor substrates. Has alpha-2,6-sialyltransferase activity toward oligosaccharides that have the Gal-beta-1,4-GlcNAc sequence at the non-reducing end of their carbohydrate groups, but it has weak or no activities toward glycoproteins and glycolipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha 2,6-ST 2; Beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; hST6Gal II; sialyltransferase; Sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6Gal II; ST6GalII
Gene Aliases: C230064G14Rik; KIAA1877; mKIAA1877; SIAT2; ST6GAL2; ST6GalII
UniProt ID: (Human) Q96JF0, (Mouse) Q76K27
Entrez Gene ID: (Human) 84620, (Mouse) 240119
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support