Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DCATAYVLL AEEEATTIVD AERLFKQALK AGETIYRQSQ QCQHQSPQHE AQLRRDTNVL VYIKRRLAMC ARKLGRIREA VKIMRDLMKE FPPLTMLNIH ENLLESLLEL QAYPDVQAVL AKYDDISLPK SAAICYTAAL LKTRTVSEKF SPETASRRGL STAEINAVEA IHRAVEFNPH VPKYLLEMKS LILPPEHILK RGDSEAIAYA FFHLQHWKRI EGALNLLQCT WEGTFRMIPY PLEKGHLFYP YPSCTETADR ELLPTFHHVS VYPKKELPLF IHFTAGFCSS TAMIAILTHQ FPEIMGIFAK AVLGLWCPQP WASSGFEENT QDLKSEDLGL SSG (234-575 aa encoded by BC065501)
This gene was identified by its similarity to the ST7 tumor suppressor gene found in the chromosome 7q31 region. This gene is clustered in a tail-to-tail manner with the WNT2B gene in a chromosomal region known to be deleted and rearranged in a variety of cancers. Several transcript variants encoding many different isoforms have been described, but some have not been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ST7-related protein; Suppressor of tumorigenicity 7 protein-like; suppressor of tumorigenicity protein 7-like protein; tumor suppressor ST7 like
Gene Aliases: FAM4B; ST7L; ST7R; STLR
UniProt ID: (Human) Q8TDW2, (Mouse) Q8K4P7, (Rat) Q68FW3
Entrez Gene ID: (Human) 54879, (Mouse) 229681, (Rat) 295344
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support