Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SLISYVSLK KENIFTTPKY ASPGAPRMYM FHAGFRSQFA LKFLDPSFVP ITNSLTQELQ EKPSKWKFNR TAFLHQRQEI LQHVDVIKNF SLTKNSVRIG QLMHYDYSSH KYVFSISNNF RSLLPDVSPI MNKHYNICAV VGNSGILTGS QCGQEIDKSD FVFRCNFAPT EAFQRDVGRK TNLTTFNPSI LEKYYNNLLT IQDRNNFFLS LKKLDGAILW IPAFFFHTSA TVTRTLVDFF VEHRGQLKVQ LAWPGNIMQH VNRYWKNKHL SPKRLSTGIL MYTLASAICE EIHLYGFWPF GFDPNTREDL PYHYYDKKGT KFTTKWQESH QLPAEFQLLY RMHGEGLTKL TLSHCA (26-380 aa encoded by BC074909)
ST8SIA3 belongs to a family of sialyltransferases that form sialyl-alpha-2,8-sialyl-R linkages at the nonreducing termini of glycoconjugates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-2,8-sialyltransferase 8C; Alpha-2,8-sialyltransferase III; Alpha-N-acetylneuraminate alpha-2,8-sialyltransferase ST8SIA3; Ganglioside GD3 synthase ST8SIA3; GT3-synthase; hST8Sia III; Sia alpha 2,3Gal beta 1,4GlcNAc alpha 2,8-sialyltransferase; Sia-a2,3-Gal-b1,4-Glc-NAc-R:a2,8-sialyltransferase; sia-alpha-2,3-Gal-beta-1,4-GlcNAc-R:alpha 2,8-sialyltransferase; sialyltransferase 8 (alpha-2, 8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 2,8-sialyltransferase) C; Sialyltransferase 8 (GT3 alpha 28-sialyltransferase) C; sialyltransferase 8 C; Sialyltransferase 8C; sialyltransferase 8C (alpha2,3Galbeta1,4GlcNAcalpha 2,8-sialyltransferase); Sialyltransferase St8Sia III; sialytransferase St8Sia III; SIAT8-C; ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 3; ST8SiaIII
Gene Aliases: AI847333; SIAT8C; ST8SIA3; ST8SiaIII
UniProt ID: (Human) O43173, (Mouse) Q64689
Entrez Gene ID: (Human) 51046, (Rat) 25547, (Mouse) 20451
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support