Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RILVEESREA THGTPAALRT LRSPATAVPR ATNSTYLNEK SLQLTEKCKN LQYGIESFSN KTKGYSENDY LQIITDIQSC PWKRQAEEYA NFRAKLASCC DAVQNFVVSQ NNTPVGTNMS YEVESKKEIP IKKNIFHMFP VSQP
Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 48%.
Prefers O-glycans to N-glycans or glycolipids as acceptor substrates. The minimal acceptor substrate is the NeuAc- alpha-2,3(6)-Gal sequence at the non-reducing end of their carbohydrate groups.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-2,8-sialyltransferase 8F; alpha-2,8-sialyltransferase 8F variant 1; alpha-2,8-sialyltransferase 8F variant 2; alpha-2,8-sialyltransferase 8F variant 3; Sialyltransferase 8F; sialyltransferase 8F (alpha-2, 8-sialyltransferase); Sialyltransferase St8Sia VI; sialytransferase St8Sia VI; SIAT8-F; ST8 alpha-N-acetylneuraminate alpha-2,8-sialyltransferase 6; ST8SiaVI
Gene Aliases: SIA8F; SIAT8F; ST8SIA-VI; ST8SIA6
UniProt ID: (Human) P61647
Entrez Gene ID: (Human) 338596
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support