Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MPLFATNPFD QDVEKATSEM NTAEDWGLIL DICDKVGQSR TGPKDCLRSI MRRVNHKDPH VAMQALTLLG ACVSNCGKIF HLEVCSRDFA SEVSNVLNKG HPKVCEKLKA LMVEWTDEFK NDPQLSLISA MIKNLKEQGV TFPAIGSQAA EQAKASPALV AKDPGTVANK KEEEDLAKAI ELSLKEQRQQ STTLSTLYPS TSSLLTNHQH EGRKVRAIYD FEAAEDNELT FKAGEIITVL DDSDPNWWKG ETHQGIGLFP SNFVTAYLTA EPEMIKTEKK TVQFSDDVQV ETIEPEPEPA FIDEDKMDQL LQMLQSTDPS DDQPDLPELL HLEAMCHQMG PLIDEKLEDI DRKHSELSEL NVKVMEALSL YTKLMNEDPM YSMYAKLQNQ PGSGPTIRKP SPS
STAM (Signal transducing adaptor molecule) is an adaptor molecule involved in downstream signaling pathways mediated by growth factors and cytokines. STAM also plays an important role in endocytosis, the JAK-STAT signaling pathway, and membrane trafficking. In humans, the gene encoding STAM is located on chromosome 10.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686J2352; HSE1 homolog; Signal transducing adapter molecule 1; signal transducing adaptor molecule (SH3 domain and ITAM motif) 1; STAM-1; STAT91
Gene Aliases: STAM; STAM-1; STAM1
UniProt ID: (Human) Q92783
Entrez Gene ID: (Human) 8027
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support